Learn More
Fisherbrand™ Reduction Y with 3/4 in. x 1 in. ID - Polypropylene - QC
SDP
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.
Catalog No. 01-000-712
Description
Description
  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Specifications
Compliance Certificates Available
Non-Animal Derived Polypropylene
Tube to Tube Connector for Medical, Pharmaceutical, Industrial applications
Out of direct sunlight, >20°F <100°F
Class 7 Cleanroom Manufactured
Reduction Y
100 Pack
Natural