Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reelin C-Term Mouse anti-Mouse, Clone: 12C10, MilliporeSigma™

Mouse Monoclonal Antibody

$373.00

Specifications

Antigen Reelin C-Term
Clone 12C10
Concentration 0.5 mg/mL
Applications Immunofluorescence, Immunohistochemistry, Inhibition Assays, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN2428100
View Documents
MilliporeSigma
MABN2428100UL
100 μL
Each for $373.00
Add to cart
 
MABN242825
View Documents
MilliporeSigma
MABN242825UL
25 μL
Each for $373.00
Add to cart
 
Description

Description

Reelin C-Term Monoclonal antibody specifically detects Reelin C-Term in Mouse samples. It is validated for Immunofluorescence, Immunohistochemistry, Inhibition, Western Blotting.
Specifications

Specifications

Reelin C-Term
0.5 mg/mL
Unconjugated
Mouse
Mouse
KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Reelin.
Monoclonal
Protein G purified
12C10
Immunofluorescence, Immunohistochemistry, Inhibition Assays, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG2b in PBS without azide.
IgG2bκ
Primary
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit