Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REEP1 Mouse anti-Human, Mouse, Rat, Clone: S345-51, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527607

Catalog No. PIMA527607


Add to cart

Description

Description

REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.
Specifications

Specifications

REEP1
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q8BGH4, Q9H902
Receptor expression-enhancing protein 1, C2orf23, D6Ertd253e, rcG_56072, Reep1, SPG31
Mouse
IgG2B
100 μg
-20°C
Primary
362384, 52250, 65055
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S345-51
Unconjugated
REEP1
Liquid
REEP1
Fusion protein amino acids 111-201 of mouse REEP1
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit