Learn More
REEP1 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: S345-51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259312FR
Catalog No. NBP259312FR
Add to cart
Description
Description
REEP1 Monoclonal antibody specifically detects REEP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
REEP1
S345-51
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
REEP1
Fusion protein amino acids 111-201 of mouse REEP1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 22kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
C2orf23, chromosome 2 open reading frame 23, receptor accessory protein 1, receptor expression-enhancing protein 1, SPG31FLJ13110
Mouse
IgG2b
100 ul
Primary
65055
Human, Mouse, Rat