Learn More
REEP1 Mouse anti-Human, Mouse, Rat, FITC, Clone: S345-51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259312F
Catalog No. NBP259312F
Add to cart
Description
Description
REEP1 Monoclonal antibody specifically detects REEP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
REEP1
S345-51
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
REEP1
Fusion protein amino acids 111-201 of mouse REEP1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 22kDa.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
C2orf23, chromosome 2 open reading frame 23, receptor accessory protein 1, receptor expression-enhancing protein 1, SPG31FLJ13110
Mouse
IgG2b
100 ul
Primary
65055
Human, Mouse, Rat