Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REEP1/REEP2 Mouse anti-Human, Mouse, Rat, Clone: S326D-29, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527624

Catalog No. PIMA527624


Add to cart

Description

Description

The REEP family are transmembrane proteins that interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. REEP2 is an integral membrane protein expressed in taste cells. Unlike its counterpart REEP1, REEP2 does not increase cell surface expression to sweet receptors, but instead alters their spatial organization. REEP1 protein is a mitochondrial protein that functions to enhance the cell surface expression of odorant receptors. Mutations in the REEP1 gene cause spastic paraplegia autosomal domit type 31, a neurodegenerative disorder.
Specifications

Specifications

REEP1/REEP2
Monoclonal
0.5 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q8BGH4, Q8VCD6, Q9H902, Q9BRK0
Receptor expression-enhancing protein 2, C5orf19, SGC32445, LOC682105
Mouse
IgG1
100 μg
-20°C
Primary
52250, 225362, 362384, 51308, 65055, 682105
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S326D-29
Unconjugated
REEP2
Liquid
REEP1, REEP2
Fusion protein amino acids 111-254 (Cytoplasmic C-terminus) of mouse REEP2.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit