Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REEP2 Mouse anti-Human, Mouse, Rat, Alexa Fluor 405, Clone: S326D-29, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259314AF405
Description
REEP2 Monoclonal specifically detects REEP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
REEP2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
REEP2 | |
Fusion protein amino acids 111-254 (Cytoplasmic C-terminus) of mouse REEP2. | |
100 μL | |
51308 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S326D-29 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2 | |
Mouse | |
Protein G purified | |
Primary | |
Detects 30kDa. Cross-reacts with REEP1. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title