Learn More
REEP2 Mouse anti-Human, Mouse, Rat, Alexa Fluor 700, Clone: S326D-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259313AF700
Catalog No. NB259313700
Add to cart
Description
Description
REEP2 Monoclonal specifically detects REEP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
REEP2
S326D-2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
REEP2
Fusion protein amino acids 111-254 (cytoplasmic C-terminus) of mouse REEP2
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 30kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2
Mouse
IgG2a
100 ul
Primary
51308
Human, Mouse, Rat