Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

REEP2 Mouse anti-Human, Mouse, Rat, Clone: S326D-2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527625

Catalog No. PIMA527625



Description

Description

Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.
Specifications

Specifications

REEP2
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q8VCD6, Q9BRK0
Receptor expression-enhancing protein 2, C2orf19, SGC32445, LOC682105
Mouse
IgG2A
100 μg
-20°C
Primary
225362, 51308, 682105
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S326D-2
Unconjugated
REEP2
Liquid
REEP2
Fusion protein amino acids 111-254 (cytoplasmic C-terminus) of mouse REEP2
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit