Learn More
REEP2 Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: S326D-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259313R
Catalog No. NBP259313R
Add to cart
Description
Description
REEP2 Monoclonal antibody specifically detects REEP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
REEP2
S326D-2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
REEP2
Fusion protein amino acids 111-254 (cytoplasmic C-terminus) of mouse REEP2
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 30kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2
Mouse
IgG2a
100 ul
Primary
51308
Human, Mouse, Rat