Learn More
REEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156841AP
Catalog No. 50-173-0403
Add to cart
Description
Description
Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.
Specifications
Specifications
REEP2
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8VCD6, Q9BRK0
C5orf19, receptor accessory protein 2, REEP2, SGC32445
Rabbit
IgG
150 μL
-20°C
Primary
225362, 51308, 682105
Flow Cytometry, Western Blot
Unconjugated
REEP2
Liquid
REEP2
REEP2 Fusion Protein Ag8186
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat