Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reference Color Standard No.11, ASTM D 848, Ricca Chemical
SDP

Catalog Number r6606100500
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
7778-50-9, 7789-00-6, 7732-18-5
Cr2K2O7
294.182
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
7778-50-9, 7789-00-6, 7732-18-5
Cr2K2O7
294.182
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Packaging Amber Glass Bottle
Quantity 500 mL
Color Colorless
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Reference Color Standard No. 11, ASTM D 848
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit