Learn More
Brady™ Reflective Quik-Lite™ Ten Packs - Printed Letter Upper Case: G
Identify equipment, bins and areas of warehouse required by facility safety and maintenance. Brady™ Reflective Quik-Lite™ Ten Packs can be used to mark tool cribs, bins, shelves, poles, panels, switches and equipment.
Supplier:  Brady™ 9740G
 View more versions of this product
Catalog No. 19-108-788
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Made of B-957 reflective material (Vinyl)
  • With permanent, aggressive adhesive grips firmly to most clean, dry surfaces, indoors and outdoors
Specifications
Specifications
Reflective: Engineer-Grade
White
Reflective Vinyl Sheeting (B-957)
G
10/Pk.
2 in.
Acrylic, Permanent
Die-cut
Reflective Quik-Lite, Printed Letter, Upper Case
Labeling in Low Light Conditions; Safety and Compliance ID
0.013 in.
Product Suggestions
Product Suggestions