Learn More
Brady™ Reflective Quik-Lite™ Ten Packs - Printed Letter Upper Case: I
Identify equipment, bins and areas of warehouse required by facility safety and maintenance. Brady™ Reflective Quik-Lite™ Ten Packs can be used to mark tool cribs, bins, shelves, poles, panels, switches and equipment.
Manufacturer:  Brady™ 9720I
 View more versions of this product
Catalog No. 19-108-710
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Made of B-957 reflective material (Vinyl)
  • With permanent, aggressive adhesive grips firmly to most clean, dry surfaces, indoors and outdoors
Specifications
Specifications
Reflective Quik-Lite, Printed Letter, Upper Case
White
Reflective: Engineer-Grade
Labeling in Low Light Conditions; Safety and Compliance ID
Reflective Vinyl Sheeting (B-957)
Die-cut
Acrylic, Permanent
I
1 in.
0.013 in.
10/Pk.