Learn More
Brady™ Reflective Quik-Lite™ Ten Packs - Printed Symbol: ARROW
Identify equipment, bins and areas of warehouse required by facility safety and maintenance. Brady™ Reflective Quik-Lite™ Ten Packs can be used to mark tool cribs, bins, shelves, poles, panels, switches and equipment.
Manufacturer:  Brady™ 9750ARO
 View more versions of this product
Catalog No. 19-108-822
  Check Availability
Add to cart
Specifications
Description
  • Made of B-957 reflective material (Vinyl)
  • With permanent, aggressive adhesive grips firmly to most clean, dry surfaces, indoors and outdoors
Description & Specifications
Specifications
Acrylic,Permanent
ARROW SYMBOL
3 in.
0.013 in.
Reflective Vinyl Sheeting (B-957)
10/Pk.
White
Reflective: Engineer-Grade
Labeling in Low Light Conditions; Safety and Compliance ID
Die-cut on cards
Reflective Quik-Lite, Printed Symbol