Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rotor Adapters
For 6 × 85mL fixed-angle rotors
$206.00 - $239.07
Specifications
For Use With (Equipment) | Eppendorf™ special rotor for 5804/5804R/5810/5810R microcentrifuge |
---|
Catalog Number | Mfr. No. | Bore Size (Metric) | No. of Places | Tube Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bore Size (Metric) | No. of Places | Tube Size | Price | Quantity & Availability | |||
05-400-85
|
Eppendorf™
022637240 |
2.6 cm | 1 | 20 to 30 mL |
Pack of 2 for $206.00
|
|
05-400-82
|
Eppendorf™
022637215 |
1.1 cm | 4 | 1.5 to 2 mL |
Pack of 2 for $239.07
|
|
05-400-83
|
Eppendorf™
022637223 |
1.6 cm | 2 | 4 to 10 mL |
Pack of 2 for $239.07
|
|
Description
For use with Standard and Vacutainer conical and micro test tubes.
Specifications
Eppendorf™ special rotor for 5804/5804R/5810/5810R microcentrifuge |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title