Learn More
Saf T Pak Inc STP-311 - Outer Box for STP-310 Shipping System, (UN2814, UN 2900 and UN1845)
Supplier: Saf T Pak Inc STP311
STP-311 - Outer Box for STP-310 Shipping System, (UN2814, UN 2900 and UN1845) 10/Case Outer Box – OD 15.5 x 13.75 x 12.75”, pre-printed with: Class 6 Infectious Substance Label, UN2814 Marking, Class 9 Hazard Label, & Dry Ice Marking (UN 1845)