Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Reg1B Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
$1219.00
Specifications
Assay Range | 3.91 pg/mL |
---|---|
Assay Sensitivity | 3.91-250 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009745
|
Novus Biologicals™
NBP27935315PLATES |
15 Plates |
Each of 1 for $1,219.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Reg1B.Specifications
3.91 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2 | |
REG1B | |
RUO | |
Reg1B |
3.91-250 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Reg1B. | |
Sandwich ELISA | |
5968 | |
Rabbit | |
Storage is content dependent. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title