Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg2 Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0312-5D45, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212429AF350

Catalog No. NP212429A30


Add to cart

Description

Description

Reg2 Monoclonal antibody specifically detects Reg2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Reg2
RM0312-5D45
Western Blot
Conjugated, Purified
REG1B
Mouse
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
regenerating islet-derived 2, RegII
Rat
IgG2
0.1 mL
Primary
5968.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit