Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reg3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$206.50 - $427.50
Specifications
Antigen | Reg3A |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 2.5-5 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224763SS
![]() |
Novus Biologicals
NBP224763SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224763
![]() |
Novus Biologicals
NBP224763 |
0.1 mg |
Each for $427.50
|
|
|||||
Description
Reg3A Polyclonal specifically detects Reg3A in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Reg3A | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 2.5-5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
5068 | |
A portion of amino acids 1-50 of human REG3A was used as the immunogen for this antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Primate | |
hepatocarcinoma-intestine-pancreas, HIPpancreatitis-associated protein, Human proislet peptide, pancreatic beta cell growth factor, Pancreatitis-associated protein 1, PAP homologous protein, PAP1FLJ18565, PAPPAP-H, PBCGF, proliferation-inducing protein 34, proliferation-inducing protein 42, Reg III-alpha, REG3, REG-3-alpha, regenerating islet-derived 3 alpha, regenerating islet-derived protein 3-alpha, Regenerating islet-derived protein III-alpha, REG-III | |
REG3A | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 89% homologous in monkey and 73% homologous in cow and dog. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title