Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg3B Rabbit anti-Mouse, Alexa Fluor 405, Clone: 5, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290800AF405

Catalog No. NB041040


Add to cart

Description

Description

Reg3B Monoclonal antibody specifically detects Reg3B in Mouse samples. It is validated for Western Blot
Specifications

Specifications

Reg3B
5
Western Blot
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor 405
50mM Sodium Borate
PAP1, Reg3B
Recombinant Mouse Reg3A Protein
Protein A purified
RUO
Primary
18489
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit