Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REG3G Monoclonal Antibody (C15), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $458.50
Specifications
Antigen | REG3G |
---|---|
Clone | C15 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA550940
|
Invitrogen™
MA550940 |
20 μL |
Each for $195.80
|
|
|||||
FEMA550939
|
Invitrogen™
MA550939 |
200 μL |
Each for $458.50
|
|
|||||
Description
REG3G might be a stress protein involved in the control of bacterial proliferation.Specifications
REG3G | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
AI449515; generating islet-derived, mouse homolog 3 gamma; LPPM429; Pancreatitis-associated protein 1B; pancreatitis-associated protein 3; Pancreatitis-associated protein IB; PAP IB; PAP1B; PAP-1B; Pap3; PAPIB; PAPIII; REG III; Reg III-gamma; REG3G; REG3gamma; REG-3gamma; REG-3-gamma; regenerating family member 3 gamma; regenerating gene III; regenerating islet-derived 3 gamma; regenerating islet-derived protein 3 gamma; regenerating islet-derived protein 3-gamma; Regenerating islet-derived protein 3-gamma 15 kDa form; Regenerating islet-derived protein 3-gamma 16.5 kDa form; Regenerating islet-derived protein III-gamma; REG-III; RegIII (gamma); UNQ429; UNQ429/PRO162 | |
REG3G | |
IgG2a κ | |
Protein A/G | |
REG3G |
C15 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q6UW15 | |
130120 | |
Recombinant Human REG3g protein, Ser39-Asp175 (Accession #Q6UW15). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title