Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ REG3G Monoclonal Antibody (D9)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | REG3G |
---|---|
Clone | D9 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541666
|
Invitrogen™
MA541666 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543611
|
Invitrogen™
MA543611 |
20 μL |
Each for $243.00
|
|
|||||
Description
REG3G Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
REG3G might be a stress protein involved in the control of bacterial proliferation.Specifications
REG3G | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q6UW15 | |
130120 | |
Recombinant protein Regenerating Islet Derived Protein 3 Gamma. The antigen corresponds to amino acid range 39-175 of the target protein. | |
Antibody |
D9 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
REG3G | |
AI449515; generating islet-derived, mouse homolog 3 gamma; LPPM429; Pancreatitis-associated protein 1B; pancreatitis-associated protein 3; Pancreatitis-associated protein IB; PAP IB; PAP1B; PAP-1B; Pap3; PAPIB; PAPIII; REG III; Reg III-gamma; REG3G; REG3gamma; REG-3gamma; REG-3-gamma; regenerating family member 3 gamma; regenerating gene III; regenerating islet-derived 3 gamma; regenerating islet-derived protein 3 gamma; regenerating islet-derived protein 3-gamma; Regenerating islet-derived protein 3-gamma 15 kDa form; Regenerating islet-derived protein 3-gamma 16.5 kDa form; Regenerating islet-derived protein III-gamma; REG-III; RegIII (gamma); UNQ429; UNQ429/PRO162 | |
REG3G | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title