Learn More
Reg4 Mouse anti-Human, Alexa Fluor 488, Clone: MM0524-9B21, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211864AF488
Catalog No. NB211864488
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reg4 Monoclonal antibody specifically detects Reg4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Reg4
MM0524-9B21
Western Blot
Purified
REG4
Human REG4
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein
Mouse
IgG1
0.1 ml
Primary
83998