Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg4 Rabbit anti-Human, Alexa Fluor 700, Clone: 244, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289832AF700

Catalog No. NB109626


Add to cart

Description

Description

Reg4 Monoclonal specifically detects Reg4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Reg4
244
50 mM Sodium Borate
Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein
This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg4 (NP_001152824.1; Met 1-Pro 158).
Protein A purified
RUO
Primary
83998
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 700
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit