Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Uniflo Syringe Filter, Regenerated Cellulose Membrane
Offers broad compatibility with a wide range of aqueous and nonaqueous solvents. Whatman™ Uniflo Syringe Filter, Regenerated Cellulose Membrane is a disposable, hydrophilic filter unit that is mechanically stable with low protein binding, plus low extractable levels to minimize contamination.
Specifications
Material Regenerated Cellulose membrane
Housing Material Polypropylene
Membrane Hydrophilic
Sterility Non-sterile
Disposable  Yes
View More Specs
Products 7
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size No. per Pack Price Quantity & Availability  
09-927-427
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463462
25mm 0.45μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-586
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463453
25mm 0.2μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-575
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463463
25mm 0.45μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-577
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463862
13mm 0.45μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-430
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463852
13mm 0.2μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-426
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463875
13mm 0.2μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-581
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463876
13mm 0.45μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Whatman Uniflo Syringe Filters are disposable filter units designed to provide clean filtrate from small volumes up to 10mL (13mm filters) or 100mL (25mm filters).
  • Combines premium quality with economic efficiency
  • Available in a variety of membrane choices with a polypropylene overmold housing
  • A range of sizes and pore sizes to meet the filtration application needs
  • Individual ink-jet printing on the filter for easy identification
  • Packaged in a clean, compact recyclable plastic box to optimize benchtop space
Specifications
Specifications
Regenerated Cellulose membrane
Hydrophilic
 Yes
Male Slip Luer
Aqueous and nonaqueous samples
UNIFLO Syringe Filter
Polypropylene
Non-sterile
Female Luer Lock
HPLC sample filtration
Resealable, recyclable plastic boxes