Learn More
Ismatec™ Reglo Miniflex Dispensing Pump Systems
Complete systems to increase the efficiency and accuracy of filling and dispensing applications
 View more versions of this product
Catalog No. 11-475-089
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Flow range 0.16 to 365 mL/min with included L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Realize increased efficiency and throughput when compared to hand pouring, manually pipetting, or using bottle-top style dispensers
  • 5 in. (12.7 cm) capacitive TFT/RGB touch screen puts the intuitive interface at your fingertips
  • Dispense by volume or time -- anti-drip and speed ramping available in Volume dispense mode
  • Flexible operational control via screen, MasterflexLive, or analog remote control
  • Pump lets you save and retrieve commonly used dispense protocols for rapid set up
Specifications
Specifications
0.16 to 365 mL/min
90 to 260 V, 50/60 Hz
Dependent on tubing
Variable-Speed Digital
Footswitch, Stainless steel dispensing nozzle, PTFE tube weight, Dispensing stand, Rod, and Clamps
Yes
5 in. (12.7 cm) capacitive TFT/RGB touch screen
IP 31
SDS
Documents
Product Certifications
Accessories