Learn More
Selleck Chemical LLC
Regorafenib (BAY 73-4506) 10mg 755037-03-7 Fluoro-Sorafenib, Resihance, Stivarga
Manufacturer: Selleck Chemical LLC S117810MG
Regorafenib (BAY 73-4506, Fluoro-Sorafenib, Resihance, Stivarga) is a multi-target inhibitor for VEGFR1, VEGFR2, VEGFR3, PDGFRβ, Kit (c-Kit), RET (c-RET) and Raf-1 with IC50 of 13 nM/4.2 nM/46 nM, 22 nM, 7 nM, 1.5 nM and 2.5 nM in cell-free assays, respectively. Regorafenib induces autophagy.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.