Learn More
PIP™ Regular Grade Top Grain Cowhide Leather Palm Glove
PIP™ Regular Grade Top Grain Cowhide Leather Palm Glove is ideal for pipe, brick and other material handling, construction, farming and general maintenance.
 View more versions of this product
Catalog No. 19-048-666B
Add to cart
Description
Description
  • Cowhide leather construction provides comfort, durability, excellent abrasion resistance, and breathability
  • Cotton lined palm for extra comfort
  • Gunn cut pattern delivers excellent durability, wear and comfort, especially for heavier gloves
  • Wing thumb increases comfort in open-handed work
  • Fabric back provides breathability and flexibility
  • Leather knuckle strap and finger tips for extra protection
  • Elastic closure sewn inside glove for safety and comfort
  • Gauntlet cuff for added forearm protection and allows for easy on and off
  • Color coded hems for easy size identification
Specifications
Specifications
Gauntlet
Cowhide Leather
Blue
Cotton lined palm
Medium
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Warning - Prop 65 - Cancer and Reproductive Harm