Learn More
Reichert™ AR700™ and AR70™ Temperature Controlled Automatic Refractometers
Accurate readings of Brix, Refractive Index, product concentration measurement, purity determination, and chemical identification analysis at specified temperatures
Manufacturer:  Reichert™ 13107000
 View more versions of this product
Catalog No. 13-975-118
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
UL, CE
LCD
4.5 in.
47/63Hz
RS232C
430mm
0.00001
0.01°C
17 lb.
10.5 in.
0.00005
10 to 100°C
17 x 10.5 x 4.5 in. (430 x 270 x 110mm)
110/220V 47/63Hz
110mm
NIST certified Oils
17 in.
1.33 to 1.6
Transparent, translucent and opaque liquids
110/220V
7700g
270mm
50 to 212°F