Reichert™ Industrial Fluid Testing Refractometers
Designed specifically for checking ratio or concentration of water-soluble industrial fluids
Manufacturer: Reichert™ 13851000
 View more versions of this product
Catalog No. 13-975-112
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Includes Durable Plastic carrying case, sample application pipets, calibration adjustment wrench
    Increments 0.2%
    Refractive Index 1.33 to 1.35
    Temperature Compensation 15 to 38°C
    Calibration Distilled Water
    Accuracy ±0.2
    Scale Range 0 to 10%
    For Use With (Application) Ideal for applications including Metalworking coolants and lubricants, heat treating fluids, hydraulic fluids, firefighting Foam and plating baths
  • Description & Specifications
Related Products
Product Content Feedback