Reichert™ Rhino Ultra-Durable Handheld Refractometers

Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant

Manufacturer: Reichert™ 137531L0

 View more versions of this product

Catalog No. 13-975-110

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance IIP67
    Increments 0.25%
    Refractive Index 1.333 to 1.42
    Accuracy ±0.25
    Scale Range 0 to 50% Brix
    For Use With (Application) Food and beverage, chemical, pharmaceutical and industrial cutting fluids

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit