Learn More
Relaxin R1/LGR7 Mouse anti-Human, Mouse, Rat, Alexa Fluor 647, Clone: 933344, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8898R100UG
Catalog No. FAB8898R100
Add to cart
This item is not returnable. View return policy
Description
Description
Relaxin R1/LGR7 Monoclonal antibody specifically detects Relaxin R1/LGR7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
Relaxin R1/LGR7
Monoclonal
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RelaxinR1, RXFP1, RXFPR1
Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R1 Met1-Ser398 Accession # Q9HBX9
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Relaxin R1 in direct ELISAs and detects human, mouse, and rat Relaxin R1 in Western blots.
Flow Cytometry
933344
Flow Cytometry 0.25-1 ug/10^6 cells
Q9HBX9
Mouse
IgG1
RUO
Primary
59350
Human, Mouse, Rat
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)