Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RelB Antibody (9E4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | RelB |
---|---|
Clone | 9E4 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
RelB Monoclonal antibody specifically detects RelB in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
RelB | |
Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
I-Rel, IREL, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, v-rel reticuloendotheliosis viral oncogene homolog B | |
A synthesized peptide derived from Human RelB [UniProt Q01201] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9E4 | |
Flow Cytometry, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5971 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title