Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RELB Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30535
Description
Rabbit polyclonal antibody raised against recombinant human RELB.
- High specificity
- High reactivity
Specifications
RELB | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human RELB. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
I-REL/IREL | |
Rabbit | |
Antigen affinity purification | |
RUO | |
5971 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500), Immunofluorescence (1-4 ug/mL), The optimal working dilution should be determined by the end user. | |
Q01201 | |
RELB | |
Recombinant protein corresponding to human RELB. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction