Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RELL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen RELL2
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096263
View Documents
Novus Biologicals
NBP305042100UL
100 μg Each for $320.71
Add to cart
 
NB096264
View Documents
Novus Biologicals
NBP30504220UL
20 μg Each for $138.37
Add to cart
 
Description

Description

RELL2 Polyclonal antibody specifically detects RELL2 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

RELL2
Unconjugated
Rabbit
Human
C5orf16, chromosome 5 open reading frame 16, FLJ90583, receptor expressed in lymphoid tissues like 2, RELT-like 2, RELT-like protein 2
Recombinant fusion protein containing a sequence corresponding to amino acids 44-303 of human RELL2 (NP_776189.3).
Polyclonal
Affinity purified
Immunohistochemistry
Purified
RUO
PBS with 50% glycerol, pH7.3.
285613
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit