Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RELM beta, Rat anti-Mouse, Clone: MRB 46L, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280033100UG
Description
RELM beta Monoclonal specifically detects RELM beta in Mouse samples. It is validated for Western Blot, ELISA.Specifications
RELM beta | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA | |
C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Recognizes mouse RELM beta. Detects a band of ∽10kDa by Western blot. Does not cross-react with human RELM beta. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA | |
MRB 46L | |
Unconjugated | |
0.2 um-filtered solution in PBS, pH 7.4 | |
RETNLB | |
Recombinant mouse RELM beta. | |
100 μg | |
Cancer | |
84666 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction