Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RELM beta, Rat anti-Mouse, Clone: MRB 46L, Novus Biologicals™

Rat Monoclonal Antibody

$240.50 - $314.00

Specifications

Antigen RELM beta
Clone MRB 46L
Applications Western Blot, ELISA
Host Species Rat
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013143
View Documents
Novus Biologicals
NBP280033100UG
100 ug
Each for $314.00
Add to cart
 
NB013144
View Documents
Novus Biologicals
NBP28003350UG
50 ug
Each for $240.50
Add to cart
 
Description

Description

RELM beta Monoclonal antibody specifically detects RELM beta in Mouse Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

RELM beta
Western Blot, ELISA
Mouse
Primary
MRB 46L
Rat
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit