Learn More
Remacemide hydrochloride, Tocris
NMDA antagonist; blocks ion channel and allosteric modulatory site
Manufacturer:  R&D Systems 1622/50
Quantity 50mg
 View more versions of this product
Catalog No. 16-225-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
304.82
Remacemide hydrochloride
C17H20N2O·HCl
2-Amino-N-(1-methyl-1, 2-diphenylethyl)acetamide hydrochloride
Specifications
111686-79-4
C17H20N2O·HCl
2-Amino-N-(1-methyl-1, 2-diphenylethyl)acetamide hydrochloride
Remacemide hydrochloride
>99%
304.82
50mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature