Learn More
Thermo Scientific™ Remel™ Acetamide Agar Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Acetamide Agar is used for differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa.
$263.55 - $931.88
Specifications
Description Acetamide Agar
Format 15mm x 103mm Tube
View More Specs
Products 2
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
R060010
Accessories View Documents
Thermo Scientific™
R060010
20/Pk. Case of 100 for $931.88
Add to cart
R060012 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R060012
100/Pk. Pack of 20 for $263.55
Add to cart
Description
Description
Thermo Scientific™ Remel Acetamide Agar is used for differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Acetamide Agar
15mm x 103mm Tube