Learn More
Thermo Scientific™ Remel™ Acetamide Agar
Thermo Scientific™ Remel Acetamide Agar is used for differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa.
Supplier: Thermo Scientific™ R060012
Description
Thermo Scientific™ Remel Acetamide Agar is used for differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa.
Specifications
Acetamide Agar | |
15mm x 103mm Tube | |
Agar | |
100/Pk. |