Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Supplier: Thermo Scientific™ R30166301
This item is not returnable.
View return policy
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
Agglutinating Sera, Haemophilus influenzae Type D | |
H. influenzae | |
Agglutinating Sera |