Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Supplier: Thermo Scientific™ R30163401
This item is not returnable.
View return policy
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Positive | |
Yes | |
Agglutinating Sera | |
2 mL |
Salmonella H (1, 5) | |
Salmonella H Antigen | |
2 mL/Vial | |
Salmonella |