Learn More
Thermo Scientific™ Remel™ Ammonium Hydroxide Concentrate
Improve the recovery of dimorphic pathogenic fungi when used with Yeast Extract Phosphate Agar.
Supplier: Thermo Scientific™ R21195
Description
Improve the recovery of dimorphic pathogenic fungi with Thermo Scientific™ Remel™ Ammonium Hydroxide Concentrate.The use of ammonium hydroxide concentrate with Yeast Extract Phosphate Agar helps in primary recovery of dimorphic pathogenic fungi as demonstrated by Smith and Goodman1. Ammonium hydroxide is a selective agent that improves recovery of dimorphic pathogens2. One drop of concentrated ammonium hydroxide (NH4OH) is added to inoculated agar plate to decontaminate the sample3.
Ammonium Hydroxide Concentrate reagent is recommended for use with Yeast Extract Phosphate Agar Part Nos. R01994, R09979, R10230 to enhance recovery of dimorphic pathogenic fungi.Selective: Inhibits bacteria, some yeasts and saprophytic fungi.Improves recovery of slow-growing, dimorphic pathogens.Easy to interpret: Pink color develops within 20 minutes when one drop of Ammonium Hydroxide Concentrate is added to the surface of inoculated Yeast Extract Phosphate Agar plate.Ammonium Hydroxide Concentrate contains 1N Ammonium Hydroxide supplied as a 25mL bottle Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.Specifications
Ammonium Hydroxide Concentrate | |
Ammonium Hydroxide | |
25 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.