Learn More
Thermo Scientific™ Remel™ Anaerobic Reducible Blood Agar Available on GSA/VA Contract for Federal Government customers only.
Blood agar (Columbia Blood Agar base) is used for the primary cultivation and isolation of fastidious anaerobic microorganisms.
Manufacturer:  Thermo Scientific™ R01061
 View more versions of this product
Catalog No. R01061
Add to cart
Description
Description
Thermo Scientific™ Remel Anaerobic Reducible Blood Agar (Columbia Blood Agar base) is used for the primary cultivation and isolation of fastidious anaerobic microorganisms.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Anaerobic Reducible Blood Agar
Agar
10mm x 100mm Spacesaver Plate
15/Pk.