Learn More
Thermo Scientific™ Anaerobic Reducible LKV Blood Agar
Cultivate and isolate fastidious anaerobic bacteria with this selective, ready-to-use solid medium.
$161.00
Catalog Number | Mfr. No. | dimFormat | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimFormat | Price | Quantity & Availability | |||||
R01068
|
Thermo Scientific™
R01068 |
Pack of 10 for $161.00
|
|
||||||
R01069
|
Thermo Scientific™
R01069 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||||
Description
Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible LKV Blood Agar. Adding reducing agents lowers the oxidation-reduction potential of the medium and improves the recovery of anaerobic organisms1. Kanamycin and vancomycin are added as selective agents for recovery of gram-negative anaerobes, especially Bacteriodes spp2.
Anaerobic Reducible Blood Agar is a solid media recommended for use in qualitative procedures for the primary isolation and cultivation of fastidious anaerobic bacteria.
- Selective: Kanamycin and vancomycin antibiotics inhibit gram-positive organisms and facultative anaerobic bacteria.
- Easy-to-detect: Laked blood is added to enhance pigment formation by anaerobic bacteria.
- Ready-to-use: Convenience of prepared media.
Anaerobic Reducible Blood Agar is a nutritious medium containing peptone, hemin, vitamin K and sheep blood, which supply anaerobic organisms with required growth factors. Colistin and nalidixic inhibit the growth of most gram-negative bacilli. Kanamycin and vancomycin inhibit gram-positive organisms and facultative anaerobic bacteria. Laked blood is added to enhance pigment formation by anaerobic bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Anaerobic Reducible LKV Blood Agar | |
Agar |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.