Learn More
Thermo Scientific™ Remel™ Andrade's Broth
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
Manufacturer:  Thermo Scientific™ R060130
 View more versions of this product
Catalog No. R060130
Add to cart
Description
Description
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
Specifications
Specifications
Andrade Broth with 1% Inositol
20 x 5mL
20/Pk.
Broth
15mm x 103mm Tube
SDS
Documents
Accessories