Learn More
Thermo Scientific™ Remel™ Andrade's Broth Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
Manufacturer:  Thermo Scientific™ R060138
 View more versions of this product
Catalog No. R060138
Add to cart
Description
Description
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Andrade Broth with 1% Lactose
20 x 5mL
20/Pk.
Broth
15mm x 103mm Tube
SDS
Documents
Accessories