Learn More
Thermo Scientific™ Remel™ Andrade's Broth
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
Supplier:  Thermo Scientific™ R060174
 View more versions of this product
Catalog No. R060174
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Andrade Broth with 1% Trehalose
15mm x 103mm Tube
Microbiology Media
20 x 5 mL
SDS
Documents
Documents
Product Certifications
Accessories