Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
Supplier: Thermo Scientific™ R08070
This item is not returnable.
View return policy
Description
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
- Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
- Gentamicin inhibits some gram-negative bacteria
Specifications
BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin | |
Tube | |
Agar | |
100/Pk. |